Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01447000_m1
          See other TRPC1 GE Assays ›
          Gene Symbol:
          Trpc1
          Gene Name:
          transient receptor potential cation channel, subfamily C, member 1
          Gene Aliases:
          Trrp1
          Chromosome Location:
          Chr.8: 103503982 - 103554862 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01447000_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Trpc1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053558.1 NP_446010.1 11 - 12 2073 82
          XM_008766568.1 XP_008764790.1 12 - 13 2656 82
          XM_008766569.1 XP_008764791.1 11 - 12 1949 82
          XM_008766570.1 XP_008764792.1 10 - 11 1943 82
          XM_008766571.1 XP_008764793.1 10 - 11 1847 82
          GenBank mRNA AF061266.1 - 11 - 12 2127 82
          DQ839447.1 - 12 - 13 2154 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800003159.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Ion Channel 3' Most Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Trpc1

          Panther Classification:

          Molecular Function -

          ion channel transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          saliva secretion
          positive regulation of release of sequestered calcium ion into cytosol
          regulation of cytosolic calcium ion concentration
          response to calcium ion
          calcium ion transmembrane transport
          cellular response to hypoxia
          cation channel activity
          calcium channel activity
          store-operated calcium channel activity
          ion channel binding
          inositol 1,4,5 trisphosphate binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P242924 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P093502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345584 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 62255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371968 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 522249 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 129653 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287860 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 619383 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 771876 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 899928 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642154 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779183 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 578251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 815962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548786 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 524567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869561 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 734238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 585738 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721399 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550887 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 236147 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 842263 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 868866 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 942078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459518 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53252 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 153877 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 104103 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 20666 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 154023 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7239 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P093502 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P242924 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P242924 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P093502 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P242924 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P093502 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P242924 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701275404 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36445 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68594 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33325 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 44941 - -
          Agilent Rat cDNA Microarray (G4105A) 701275404 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10919343 - -
          Affymetrix Rat Gene 1.0 ST Array 10919345 - -
          Affymetrix Rat Gene 1.0 ST Array 10919346 - -
          Affymetrix Rat Gene 1.0 ST Array 10919340 - -
          Affymetrix Rat Gene 1.0 ST Array 10919344 - -
          Affymetrix Rat Gene 1.0 ST Array 10919350 - -
          Affymetrix Rat Gene 1.0 ST Array 10919348 - -
          Affymetrix Rat Gene 1.0 ST Array 10919352 - -
          Affymetrix Rat Gene 1.0 ST Array 10919349 - -
          Affymetrix Rat Gene 1.0 ST Array 10919341 - -
          Affymetrix Rat Gene 1.0 ST Array 10919351 - -
          Affymetrix Rat Gene 1.0 ST Array 10919347 - -
          Affymetrix Rat Gene 1.0 ST Array 10919353 - -
          Affymetrix Rat Gene 1.0 ST Array 10919342 - -
          Affymetrix Rat Gene 1.1 ST Array 10919339 - -
          Rat Genome 230 2.0 Array 1370411_PM_at - -
          Rat Genome 230 2.0 Array 1370411_at - -
          Affymetrix HT Rat Focus Array 1370411_PM_at - -
          Affymetrix HT Rat Focus Array 1370411_at - -
          Affymetrix Rat Clariom S Assay PSR0800025213.rn.1 TC0800003159.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800025214.rn.1 TC0800003159.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800025215.rn.1 TC0800003159.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800025212.rn.1 TC0800003159.rn.1 RTA TC
          Rat Expression Set 230 1370411_at - -
          Rat Expression Set 230 1370411_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10919339 - -
          Rat Genome U34 Set AF061266_g_at - -
          Rat Genome U34 Set AF061266_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22423129 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline