Search
Search
| Gene Symbol: Birc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023987.3 | NP_076477.3 | 5 - 6 | 1264 | 78 | |
| XM_006242499.2 | XP_006242561.1 | 4 - 5 | 3227 | 78 | ||
| XM_006242500.2 | XP_006242562.1 | 5 - 6 | 1997 | 78 | ||
| XM_006242502.2 | XP_006242564.1 | 4 - 5 | 1496 | 78 | ||
| XM_006242503.2 | XP_006242565.1 | 4 - 5 | 1455 | 78 | ||
| GenBank mRNA | AF183430.1 | - | 3 - 4 | 1032 | 78 | |
| BC083555.1 | - | 5 - 6 | 1264 | 7112819 | 78 | |
Set Membership: |
|