Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01447966_m1
          See other SMCO4 GE Assays ›
          Gene Symbol:
          Smco4
          Gene Name:
          single-pass membrane protein with coiled-coil domains 4
          Gene Aliases:
          RGD1309410
          Chromosome Location:
          Chr.8: 14060394 - 14079972 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01447966_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Smco4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001134584.1 NP_001128056.1 1 - 2 213 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800000125.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P809733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 575431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70581 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 777844 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 94645 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47098 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 626466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 237100 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304030 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 951067 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 832972 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13102 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 154431 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P809733 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P809733 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P809733 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P809733 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10908109 - -
          Affymetrix Rat Gene 1.0 ST Array 10908110 - -
          Affymetrix Rat Gene 1.1 ST Array 10908108 - -
          Rat Genome 230 2.0 Array 1372843_PM_at - -
          Rat Genome 230 2.0 Array 1372843_at - -
          Affymetrix HT Rat Focus Array 1372843_PM_at - -
          Affymetrix HT Rat Focus Array 1372843_at - -
          Affymetrix Rat Clariom S Assay PSR0800000782.rn.1 TC0800000125.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800000783.rn.1 TC0800000125.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800000779.rn.1 TC0800000125.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800000780.rn.1 TC0800000125.rn.1 RTA TC
          Rat Expression Set 230 1372843_at - -
          Rat Expression Set 230 1372843_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10908108 - -
          Rat Genome U34 Set rc_AI233276_at - -
          Rat Genome U34 Set rc_AI172530_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20718175 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373690 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline