Search
Search
| Gene Symbol: Erp44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008317.1 | NP_001008318.1 | 6 - 7 | 662 | 66 | |
| GenBank mRNA | AY158662.1 | - | 6 - 7 | 662 | 66 | |
| FQ227188.1 | - | 6 - 7 | 722 | 66 | ||
Set Membership: |
|