Search
Search

Atp6v1h| Gene Symbol: Atp6v1h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006237801.1 | XP_006237863.1 | 1 - 2 | 183 | 90 | |
| XM_006237804.1 | XP_006237866.1 | 1 - 2 | 183 | 90 | ||
| GenBank mRNA | FQ212744.1 | - | 1 - 2 | 162 | 90 | |
Set Membership: |
|