Search
Search
| Gene Symbol: Tram1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007701.1 | NP_001007702.1 | 902 | 159 | ||
| GenBank mRNA | AY387055.1 | - | 785 | 159 | ||
| BC083851.1 | - | 902 | 7123274 | 159 | ||
| FQ223598.1 | - | 774 | 159 | |||
Set Membership: |
|