Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01450016_m1
          See other LRRC8B GE Assays ›
          Gene Symbol:
          Lrrc8b
          Gene Name:
          leucine rich repeat containing 8 family, member B
          Gene Aliases:
          RGD1563429
          Chromosome Location:
          Chr.14: 5414872 - 5480689 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01450016_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lrrc8b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107204.2 NP_001100674.1 1 - 2 2134 85
          XM_006250557.1 XP_006250619.1 4 - 5 2445 85
          GenBank mRNA FQ212117.1 - 4 - 5 2450 85
          FQ217980.1 - 4 - 5 2400 85

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Rattus norvegicus Probe spans exons

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P20972 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P451496 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459214 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952268 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 118747 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544765 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55847 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 13710 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 891710 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 71918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 951845 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 291284 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 601372 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574717 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111543 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P20972 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P451496 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P20972 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P451496 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P20972 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P451496 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P451496 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P20972 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10775281 - -
          Affymetrix Rat Gene 1.0 ST Array 10775279 - -
          Affymetrix Rat Gene 1.0 ST Array 10775280 - -
          Affymetrix Rat Gene 1.0 ST Array 10775282 - -
          Affymetrix Rat Gene 1.1 ST Array 10775278 - -
          Rat Genome 230 2.0 Array 1378003_at - -
          Rat Genome 230 2.0 Array 1378003_PM_at - -
          Rat Expression Set 230 1378003_PM_at - -
          Rat Expression Set 230 1378003_at - -
          Affymetrix Rat Gene 1.0 ST Array 10775278 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357536 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline