Search
Search
| Gene Symbol: Coq2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044255.1 | NP_001037720.1 | 2 - 3 | 429 | 65 | |
| XM_006250675.2 | XP_006250737.1 | 2 - 3 | 461 | 65 | ||
| GenBank mRNA | BC099827.1 | - | 2 - 3 | 429 | 7934175 | 65 |
Set Membership: |
|