Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01450544_m1
          See other TMEM110 GE Assays ›
          Gene Symbol:
          Tmem110
          Gene Name:
          transmembrane protein 110
          Gene Aliases:
          -
          Chromosome Location:
          Chr.16: 6903212 - 6958786 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01450544_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem110
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_198774.2 NP_942069.1 7 - 8 856 111
          GenBank mRNA AY283801.1 - 7 - 8 879 111
          BC088092.1 - 7 - 8 856 7368117 111

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1600000085.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tmem110

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P372215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231748 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 214895 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 256090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697455 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 568179 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789273 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 967858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 133981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 704771 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 708526 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 849714 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 547195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82871 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 134091 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95782 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 83123 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911476 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692327 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 644070 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 538015 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 492383 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758440 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 401094 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 424061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 276003 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580297 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 761617 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 713061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 659666 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 683504 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 891345 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151762 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 916834 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255472 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 74944 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 794420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 249762 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 641103 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 619122 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 120200 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 155812 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 20034 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 134222 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 131881 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53020 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P372215 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P372215 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P372215 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P372215 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74533 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 96506 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27895 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18072 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 69686 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15708 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10786523 - -
          Affymetrix Rat Gene 1.0 ST Array 10786531 - -
          Affymetrix Rat Gene 1.0 ST Array 10786529 - -
          Affymetrix Rat Gene 1.0 ST Array 10786527 - -
          Affymetrix Rat Gene 1.0 ST Array 10786530 - -
          Affymetrix Rat Gene 1.0 ST Array 10786528 - -
          Affymetrix Rat Gene 1.0 ST Array 10786526 - -
          Affymetrix Rat Gene 1.0 ST Array 10786524 - -
          Affymetrix Rat Gene 1.0 ST Array 10786525 - -
          Affymetrix Rat Gene 1.1 ST Array 10786522 - -
          Rat Genome 230 2.0 Array 1390029_at - -
          Rat Genome 230 2.0 Array 1390029_PM_at - -
          Affymetrix HT Rat Focus Array 1390029_PM_at - -
          Affymetrix HT Rat Focus Array 1390029_at - -
          Affymetrix Rat Clariom S Assay PSR1600000794.rn.1 TC1600000085.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600000793.rn.1 TC1600000085.rn.1 RTA TC
          Rat Expression Set 230 1390029_at - -
          Rat Expression Set 230 1390029_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10786522 - -
          Rat Genome U34 Set rc_AA945898_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20915426 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360455 - -
          RatRef-12 Expression BeadChip ILMN_57458 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline