Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01450887_g1
          See other TNNT3 GE Assays ›
          Gene Symbol:
          Tnnt3
          Gene Name:
          troponin T type 3 (skeletal, fast)
          Gene Aliases:
          Tnt
          Chromosome Location:
          Chr.1: 215666628 - 215683628 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01450887_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tnnt3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001270664.1 NP_001257593.1 10 - 11 450 77
          NM_001270665.1 NP_001257594.1 12 - 13 483 77
          NM_001270666.1 NP_001257595.1 12 - 13 483 77
          NM_001270668.1 NP_001257597.1 10 - 11 450 77
          NM_001270670.1 NP_001257599.1 11 - 12 465 77
          NM_001270671.1 NP_001257600.1 11 - 12 465 77
          NM_001270672.1 NP_001257601.1 10 - 11 456 77
          NM_001270673.1 NP_001257602.1 11 - 12 468 77
          NM_001270675.1 NP_001257604.1 10 - 11 456 77
          NM_001270676.1 NP_001257605.1 11 - 12 468 77
          NM_001270677.1 NP_001257606.1 9 - 10 438 77
          NM_001270678.1 NP_001257607.1 11 - 12 471 77
          NM_031532.2 NP_113720.1 11 - 12 471 77
          XM_006230681.2 XP_006230743.1 10 - 11 413 77
          XM_006230683.2 XP_006230745.1 9 - 10 401 77
          XM_006230685.2 XP_006230747.1 8 - 9 383 77
          XM_006230686.2 XP_006230748.1 8 - 9 383 77
          XM_008760080.1 XP_008758302.1 13 - 14 517 77
          XM_008760081.1 XP_008758303.1 13 - 14 517 77
          XM_008760082.1 XP_008758304.1 12 - 13 467 77
          XM_008760083.1 XP_008758305.1 12 - 13 464 77
          XM_008760084.1 XP_008758306.1 11 - 12 490 77
          XM_008760085.1 XP_008758307.1 11 - 12 455 77
          XM_008760086.1 XP_008758308.1 10 - 11 437 77
          GenBank mRNA CB612473.1 - 12 - 13 432 77
          DQ062204.1 - 11 - 12 367 77
          DQ273678.1 - 11 - 12 367 77
          FQ214650.1 - 11 - 12 431 77
          FQ214722.1 - 10 - 11 418 77
          FQ214724.1 - 11 - 12 432 77
          FQ214766.1 - 10 - 11 412 77
          FQ214833.1 - 11 - 12 433 77
          FQ215030.1 - 10 - 11 412 77
          FQ215059.1 - 10 - 11 443 77
          FQ215106.1 - 10 - 11 412 77
          FQ215114.1 - 10 - 11 418 77
          FQ215252.1 - 11 - 12 430 77
          FQ215319.1 - 10 - 11 412 77
          FQ215374.1 - 11 - 12 458 77
          FQ215598.1 - 12 - 13 442 77
          FQ215694.1 - 10 - 11 450 77
          FQ215723.1 - 12 - 13 484 77
          FQ215881.1 - 12 - 13 445 77
          FQ215885.1 - 12 - 13 433 77
          FQ215897.1 - 11 - 12 458 77
          FQ215955.1 - 10 - 11 412 77
          FQ216024.1 - 7 - 8 412 77
          FQ216051.1 - 10 - 11 412 77
          FQ216059.1 - 12 - 13 440 77
          FQ216097.1 - 11 - 12 426 77
          FQ216131.1 - 10 - 11 443 77
          FQ216208.1 - 11 - 12 430 77
          FQ216324.1 - 10 - 11 412 77
          FQ216367.1 - 10 - 11 412 77
          FQ216417.1 - 11 - 12 431 77
          FQ216467.1 - 10 - 11 412 77
          FQ216480.1 - 10 - 11 418 77
          FQ216561.1 - 7 - 8 412 77
          FQ216694.1 - 11 - 12 427 77
          FQ216711.1 - 10 - 11 449 77
          FQ216750.1 - 7 - 8 303 77
          FQ216763.1 - 8 - 9 403 77
          FQ216784.1 - 10 - 11 418 77
          FQ216866.1 - 12 - 13 445 77
          FQ216925.1 - 2 - 3 174 77
          FQ217147.1 - 10 - 11 411 77
          FQ217846.1 - 12 - 13 477 77
          FQ217900.1 - 11 - 12 458 77
          FQ223840.1 - 10 - 11 412 77
          FQ223999.1 - 10 - 11 412 77
          FQ224006.1 - 10 - 11 414 77
          FQ224794.1 - 10 - 11 412 77
          FQ224813.1 - 12 - 13 445 77
          FQ224836.1 - 10 - 11 412 77
          FQ224839.1 - 11 - 12 430 77
          FQ224892.1 - 11 - 12 433 77
          FQ224966.1 - 11 - 12 416 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100002922.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tnnt3

          Panther Classification:

          Molecular Function -

          actin binding motor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          skeletal muscle contraction
          muscle contraction
          regulation of muscle contraction
          regulation of striated muscle contraction
          metabolic process
          regulation of ATPase activity
          actin binding
          tropomyosin binding
          troponin C binding
          calcium-dependent ATPase activity
          troponin I binding
          troponin T binding
          calcium-dependent protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Clariom S Assay PSR0100023492.rn.1 TC0100002922.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100023493.rn.1 TC0100002922.rn.1 RTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline