Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01451324_m1
          See other LOC499306 GE Assays ›
          Gene Symbol:
          LOC499306
          Gene Name:
          hypothetical gene supported by BC078985
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 221558093 - 221594124 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01451324_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC499306
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024291.1 NP_001019462.1 9 - 10 753 61
          GenBank mRNA BC078985.1 - 9 - 10 753 7112339 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100003095.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene LOC499306

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P098591 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 799379 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 512346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 342734 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170258 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905059 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532417 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 651444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 559058 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 812859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 315477 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 890750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 694733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164287 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701098 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 392592 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 301099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 678721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 344278 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903103 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287237 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 374034 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119647 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 14490 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 112430 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 59918 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 45116 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P098591 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P098591 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P098591 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89872 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 72896 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10713213 - -
          Affymetrix Rat Gene 1.0 ST Array 10713215 - -
          Affymetrix Rat Gene 1.0 ST Array 10713212 - -
          Affymetrix Rat Gene 1.0 ST Array 10713207 - -
          Affymetrix Rat Gene 1.0 ST Array 10713216 - -
          Affymetrix Rat Gene 1.0 ST Array 10713210 - -
          Affymetrix Rat Gene 1.0 ST Array 10713206 - -
          Affymetrix Rat Gene 1.0 ST Array 10713214 - -
          Affymetrix Rat Gene 1.0 ST Array 10713211 - -
          Affymetrix Rat Gene 1.0 ST Array 10713208 - -
          Affymetrix Rat Gene 1.0 ST Array 10713209 - -
          Affymetrix Rat Gene 1.1 ST Array 10713205 - -
          Affymetrix Rat Clariom S Assay PSR0100025129.rn.1 TC0100003095.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100025133.rn.1 TC0100003095.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100025127.rn.1 TC0100003095.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100025128.rn.1 TC0100003095.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100025136.rn.1 TC0100003095.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100025126.rn.1 TC0100003095.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100025134.rn.1 TC0100003095.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100025135.rn.1 TC0100003095.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10713205 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21243268 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1530475 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline