Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gon4l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024797.1 | NP_001019968.1 | 6 - 7 | 1280 | 95 | |
XM_006232735.2 | XP_006232797.1 | 6 - 7 | 1271 | 95 | ||
XM_006232737.2 | XP_006232799.1 | 6 - 7 | 1259 | 95 | ||
XM_006232739.2 | XP_006232801.1 | 6 - 7 | 1211 | 95 | ||
XM_006232740.2 | XP_006232802.1 | 5 - 6 | 1067 | 95 | ||
XM_006232741.2 | XP_006232803.1 | 6 - 7 | 1259 | 95 | ||
XM_008761201.1 | XP_008759423.1 | 7 - 8 | 1285 | 95 | ||
XM_008761202.1 | XP_008759424.1 | 5 - 6 | 720 | 95 | ||
GenBank mRNA | AB195689.1 | - | 5 - 6 | 1114 | 95 | |
AY724781.1 | - | 6 - 7 | 1280 | 95 | ||
AY724782.1 | - | 6 - 7 | 1280 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |