Search
Search
| Gene Symbol: Mlx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034112.1 | NP_001029284.1 | 6 - 7 | 650 | 67 | |
| XM_006247362.2 | XP_006247424.1 | 6 - 7 | 548 | 67 | ||
| GenBank mRNA | BC128702.1 | - | 6 - 7 | 684 | 7323425 | 67 |
| DQ350891.1 | - | 6 - 7 | 487 | 67 | ||
| DQ350892.1 | - | 6 - 7 | 487 | 67 | ||
| DQ350893.1 | - | 6 - 7 | 487 | 67 | ||
Set Membership: |
|