Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01453321_g1
          See other SNCG GE Assays ›
          Gene Symbol:
          Sncg
          Gene Name:
          synuclein, gamma
          Gene Aliases:
          -
          Chromosome Location:
          Chr.16: 10722110 - 10726648 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01453321_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sncg
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031688.1 NP_113876.1 4 - 5 401 72
          GenBank mRNA AY518351.1 - 5 - 6 351 72
          X86789.1 - 4 - 5 401 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1600001235.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sncg

          Panther Classification:

          Molecular Function -

          membrane trafficking regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          aggressive behavior
          chemical synaptic transmission
          peripheral nervous system development
          adult locomotory behavior
          protein secretion
          regulation of dopamine secretion
          response to cocaine
          response to drug
          regulation of neurotransmitter secretion
          synapse organization
          cellular response to hydrostatic pressure
          protein binding
          alpha-tubulin binding
          beta-tubulin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P384017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 353368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 819132 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 397989 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368938 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171975 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P384017 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P384017 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P384017 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P384017 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700370171 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 53632 - -
          Agilent Rat cDNA Microarray (G4105A) 700370171 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10790475 - -
          Affymetrix Rat Gene 1.0 ST Array 10790478 - -
          Affymetrix Rat Gene 1.0 ST Array 10790473 - -
          Affymetrix Rat Gene 1.0 ST Array 10790474 - -
          Affymetrix Rat Gene 1.0 ST Array 10790477 - -
          Affymetrix Rat Gene 1.0 ST Array 10790472 - -
          Affymetrix Rat Gene 1.1 ST Array 10790471 - -
          Rat Genome 230 2.0 Array 1398245_PM_at - -
          Rat Genome 230 2.0 Array 1398245_at - -
          Affymetrix HT Rat Focus Array 1398245_at - -
          Affymetrix HT Rat Focus Array 1398245_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1600009482.rn.1 TC1600001235.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600009483.rn.1 TC1600001235.rn.1 RTA TC
          Rat Expression Set 230 1398245_at - -
          Rat Expression Set 230 1398245_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10790471 - -
          Rat Genome U34 Set X86789_at - -
          Rat Genome U34 Set X86789_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21519739 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357880 - -
          RatRef-12 Expression BeadChip ILMN_53622 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline