Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01453533_g1
          See other CAPN7 GE Assays ›
          Gene Symbol:
          Capn7
          Gene Name:
          calpain 7
          Gene Aliases:
          -
          Chromosome Location:
          Chr.16: 7435556 - 7471808 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01453533_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Capn7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001030037.1 NP_001025208.1 20 - 21 2453 130
          XM_006252608.2 XP_006252670.2 21 - 22 2686 130
          XM_008770997.1 XP_008769219.1 20 - 21 3257 130
          GenBank mRNA BC099770.1 - 20 - 21 2453 7932056 130

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1600000099.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Protease 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Capn7

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          positive regulation of epithelial cell migration
          self proteolysis
          endopeptidase activity
          calcium-dependent cysteine-type endopeptidase activity
          MIT domain binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P619844 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 947151 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 502109 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 704436 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763326 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618329 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 37774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556773 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280378 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679932 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 222739 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 260550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 396460 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 431571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930337 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247378 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 694156 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 397147 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 613169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 755579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960928 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 585046 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 536983 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 68624 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 31328 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 2063 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P619844 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P619844 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P619844 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P21347 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74030 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2881 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19615 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10786688 - -
          Affymetrix Rat Gene 1.0 ST Array 10786695 - -
          Affymetrix Rat Gene 1.0 ST Array 10786681 - -
          Affymetrix Rat Gene 1.0 ST Array 10786684 - -
          Affymetrix Rat Gene 1.0 ST Array 10786700 - -
          Affymetrix Rat Gene 1.0 ST Array 10786687 - -
          Affymetrix Rat Gene 1.0 ST Array 10786702 - -
          Affymetrix Rat Gene 1.0 ST Array 10786691 - -
          Affymetrix Rat Gene 1.0 ST Array 10786686 - -
          Affymetrix Rat Gene 1.0 ST Array 10786692 - -
          Affymetrix Rat Gene 1.0 ST Array 10786685 - -
          Affymetrix Rat Gene 1.0 ST Array 10786683 - -
          Affymetrix Rat Gene 1.0 ST Array 10786701 - -
          Affymetrix Rat Gene 1.0 ST Array 10786694 - -
          Affymetrix Rat Gene 1.0 ST Array 10786699 - -
          Affymetrix Rat Gene 1.0 ST Array 10786690 - -
          Affymetrix Rat Gene 1.0 ST Array 10786698 - -
          Affymetrix Rat Gene 1.0 ST Array 10786682 - -
          Affymetrix Rat Gene 1.0 ST Array 10786697 - -
          Affymetrix Rat Gene 1.0 ST Array 10786689 - -
          Affymetrix Rat Gene 1.0 ST Array 10786696 - -
          Affymetrix Rat Gene 1.1 ST Array 10786680 - -
          Rat Genome 230 2.0 Array 1373111_at - -
          Rat Genome 230 2.0 Array 1373111_PM_at - -
          Affymetrix HT Rat Focus Array 1373111_at - -
          Affymetrix HT Rat Focus Array 1373111_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1600001093.rn.1 TC1600000099.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600001094.rn.1 TC1600000099.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600001092.rn.1 TC1600000099.rn.1 RTA TC
          Rat Expression Set 230 1373111_at - -
          Rat Expression Set 230 1373111_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10786680 - -
          Rat Genome U34 Set rc_AI169441_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20793639 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358048 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline