Search
Search
| Gene Symbol: Bscl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012171.1 | NP_001012171.1 | 6 - 7 | 1205 | 63 | |
| GenBank mRNA | BC089942.1 | - | 6 - 7 | 1205 | 7311394 | 63 |
| FQ220723.1 | - | 462 | 63 | |||
Set Membership: |
|