Search
Search

Mettl10| Gene Symbol: Mettl10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108504.1 | NP_001101974.1 | 2 - 3 | 253 | 112 | |
| XM_006230441.1 | XP_006230503.1 | 2 - 3 | 248 | 112 | ||
Set Membership: |
|