Search
Search
| Gene Symbol: Fank1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008347.1 | NP_001008348.1 | 3 - 4 | 387 | 124 | |
| XM_006230439.2 | XP_006230501.1 | 3 - 4 | 432 | 124 | ||
| GenBank mRNA | BC082095.1 | - | 3 - 4 | 387 | 7134234 | 124 |
Set Membership: |
|