Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01454189_m1
          See other JAK2 GE Assays ›
          Gene Symbol:
          Jak2
          Gene Name:
          Janus kinase 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 247398667 - 247457521 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01454189_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Jak2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031514.1 NP_113702.1 24 - 25 3446 86
          GenBank mRNA U13396.1 - 24 - 25 3446 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100003443.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Protein Kinase 3' Most Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Jak2

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPKK activity
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          response to oxidative stress
          signal transduction
          enzyme linked receptor protein signaling pathway
          G-protein coupled receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          JAK-STAT cascade
          tyrosine phosphorylation of STAT protein
          STAT protein import into nucleus
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          intrinsic apoptotic signaling pathway in response to oxidative stress
          hormone-mediated signaling pathway
          negative regulation of cardiac muscle cell apoptotic process
          positive regulation of cell-substrate adhesion
          positive regulation of phosphatidylinositol 3-kinase signaling
          peptidyl-tyrosine phosphorylation
          cytokine-mediated signaling pathway
          negative regulation of cell-cell adhesion
          myeloid cell differentiation
          cell differentiation
          erythrocyte differentiation
          positive regulation of cell migration
          axon regeneration
          mineralocorticoid receptor signaling pathway
          positive regulation of insulin secretion
          response to lipopolysaccharide
          positive regulation of phosphoprotein phosphatase activity
          positive regulation of interleukin-1 beta production
          positive regulation of tumor necrosis factor production
          positive regulation of protein import into nucleus, translocation
          response to hydroperoxide
          tumor necrosis factor-mediated signaling pathway
          host programmed cell death induced by symbiont
          response to tumor necrosis factor
          histone H3-Y41 phosphorylation
          intracellular signal transduction
          interleukin-12-mediated signaling pathway
          peptidyl-tyrosine autophosphorylation
          tyrosine phosphorylation of Stat3 protein
          tyrosine phosphorylation of Stat5 protein
          tyrosine phosphorylation of Stat1 protein
          positive regulation of tyrosine phosphorylation of Stat3 protein
          positive regulation of tyrosine phosphorylation of Stat5 protein
          activation of JAK2 kinase activity
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          positive regulation of DNA binding
          negative regulation of DNA binding
          negative regulation of neuron apoptotic process
          positive regulation of nitric oxide biosynthetic process
          positive regulation of cell differentiation
          negative regulation of heart contraction
          positive regulation of transcription from RNA polymerase II promoter
          response to antibiotic
          protein autophosphorylation
          platelet-derived growth factor receptor signaling pathway
          regulation of inflammatory response
          positive regulation of inflammatory response
          positive regulation of peptidyl-tyrosine phosphorylation
          positive regulation of cell activation
          positive regulation of sequence-specific DNA binding transcription factor activity
          positive regulation of nitric-oxide synthase biosynthetic process
          interferon-gamma-mediated signaling pathway
          growth hormone receptor signaling pathway
          JAK-STAT cascade involved in growth hormone signaling pathway
          positive regulation of growth hormone receptor signaling pathway
          mammary gland epithelium development
          response to interleukin-12
          cellular response to lipopolysaccharide
          cellular response to dexamethasone stimulus
          extrinsic apoptotic signaling pathway
          activation of cysteine-type endopeptidase activity involved in apoptotic signaling pathway
          positive regulation of apoptotic signaling pathway
          protein tyrosine kinase activity
          non-membrane spanning protein tyrosine kinase activity
          receptor binding
          growth hormone receptor binding
          interleukin-12 receptor binding
          protein binding
          ATP binding
          protein C-terminus binding
          protein kinase binding
          heme binding
          type 1 angiotensin receptor binding
          acetylcholine receptor binding
          histone kinase activity (H3-Y41 specific)
          SH2 domain binding
          histone binding
          phosphatidylinositol 3-kinase binding
          insulin receptor substrate binding
          peptide hormone receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P069724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 406047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 130685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959331 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 838854 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136293 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 328794 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 209183 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 51523 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323441 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 93860 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 627 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 37712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706656 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 227307 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 429128 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 305935 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 111065 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280534 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55609 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920920 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848060 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 500190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 766477 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 83165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 702707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 838043 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 422078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 364415 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781396 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369122 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 703296 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 899598 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 309765 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968896 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 285817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695148 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101908 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 137696 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 438587 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 599393 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 41185 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 40591 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 153039 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44787 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 82250 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86333 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 59608 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7969 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P069724 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P069724 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P069724 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12571 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700420836 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 101966 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 14330 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32008 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103391 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 1053 - -
          Agilent Rat cDNA Microarray (G4105A) 700420836 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10714679 - -
          Affymetrix Rat Gene 1.0 ST Array 10714676 - -
          Affymetrix Rat Gene 1.0 ST Array 10714669 - -
          Affymetrix Rat Gene 1.0 ST Array 10714668 - -
          Affymetrix Rat Gene 1.0 ST Array 10714672 - -
          Affymetrix Rat Gene 1.0 ST Array 10714670 - -
          Affymetrix Rat Gene 1.0 ST Array 10714685 - -
          Affymetrix Rat Gene 1.0 ST Array 10714680 - -
          Affymetrix Rat Gene 1.0 ST Array 10714681 - -
          Affymetrix Rat Gene 1.0 ST Array 10714684 - -
          Affymetrix Rat Gene 1.0 ST Array 10714689 - -
          Affymetrix Rat Gene 1.0 ST Array 10714688 - -
          Affymetrix Rat Gene 1.0 ST Array 10714675 - -
          Affymetrix Rat Gene 1.0 ST Array 10714686 - -
          Affymetrix Rat Gene 1.0 ST Array 10714692 - -
          Affymetrix Rat Gene 1.0 ST Array 10714677 - -
          Affymetrix Rat Gene 1.0 ST Array 10714687 - -
          Affymetrix Rat Gene 1.0 ST Array 10714674 - -
          Affymetrix Rat Gene 1.0 ST Array 10714682 - -
          Affymetrix Rat Gene 1.0 ST Array 10714683 - -
          Affymetrix Rat Gene 1.0 ST Array 10714673 - -
          Affymetrix Rat Gene 1.0 ST Array 10714671 - -
          Affymetrix Rat Gene 1.0 ST Array 10714690 - -
          Affymetrix Rat Gene 1.0 ST Array 10714693 - -
          Affymetrix Rat Gene 1.0 ST Array 10714691 - -
          Affymetrix Rat Gene 1.1 ST Array 10714667 - -
          Rat Genome 230 2.0 Array 1368856_at - -
          Rat Genome 230 2.0 Array 1368856_PM_at - -
          Rat Genome 230 2.0 Array 1380110_PM_at - -
          Rat Genome 230 2.0 Array 1380110_at - -
          Affymetrix HT Rat Focus Array 1368856_PM_at - -
          Affymetrix HT Rat Focus Array 1380110_PM_at - -
          Affymetrix HT Rat Focus Array 1380110_at - -
          Affymetrix HT Rat Focus Array 1368856_at - -
          Affymetrix Rat Clariom S Assay PSR0100027981.rn.1 TC0100003443.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100027980.rn.1 TC0100003443.rn.1 RTA TC
          Rat Expression Set 230 1368856_PM_at - -
          Rat Expression Set 230 1368856_at - -
          Rat Expression Set 230 1380110_PM_at - -
          Rat Expression Set 230 1380110_at - -
          Affymetrix Rat Gene 1.0 ST Array 10714667 - -
          Rat Genome U34 Set U13396_at - -
          Rat Genome U34 Set AJ000557cds_s_at - -
          Rat Genome U34 Set U13396_g_at - -
          Rat Genome U34 Set rc_AI145235_at - -
          Rat Genome U34 Set rc_AI105460_at - -
          Rat Genome U34 Set rc_AI229643_at - -
          Rat Genome U34 Set U13396_g_at - -
          Rat Genome U34 Set U13396_at - -
          Rat Genome U34 Set AJ000557cds_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20776171 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361170 - -
          RatRef-12 Expression BeadChip ILMN_68011 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline