Search
Search
| Gene Symbol: Phrf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139093.1 | NP_620793.1 | 11 - 12 | 1439 | 142 | |
| XM_006230510.2 | XP_006230572.1 | 12 - 13 | 1607 | 142 | ||
| XM_006230511.2 | XP_006230573.1 | 12 - 13 | 1565 | 142 | ||
| XM_006230513.2 | XP_006230575.1 | 12 - 13 | 1604 | 142 | ||
| XM_006230514.2 | XP_006230576.1 | 12 - 13 | 1607 | 142 | ||
| XM_006230515.2 | XP_006230577.1 | 12 - 13 | 1607 | 142 | ||
| XM_006230516.2 | XP_006230578.1 | 12 - 13 | 1607 | 142 | ||
| XM_006230517.2 | XP_006230579.1 | 12 - 13 | 1604 | 142 | ||
| GenBank mRNA | U49057.1 | - | 11 - 12 | 1439 | 142 | |
Set Membership: |
|