Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01454280_m1
          See other ANXA1 GE Assays ›
          Gene Symbol:
          Anxa1
          Gene Name:
          annexin A1
          Gene Aliases:
          Anx1, p35
          Chromosome Location:
          Chr.1: 237893983 - 237921165 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01454280_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Anxa1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012904.2 NP_037036.1 11 - 12 921 98
          XM_006231159.1 XP_006231221.1 12 - 13 1089 98
          XM_008760295.1 XP_008758517.1 11 - 12 1008 98
          GenBank mRNA BC061710.1 - 11 - 12 919 5598112 98
          FQ215033.1 - 11 - 12 936 98
          FQ216134.1 - 11 - 12 936 98
          FQ228716.1 - 11 - 12 936 98
          FQ229071.1 - 11 - 12 936 98
          FQ229198.1 - 10 - 11 857 98
          M19967.1 - 10 - 11 888 98
          Y00446.1 - 11 - 12 922 98

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100007387.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Anxa1

          Panther Classification:

          Molecular Function -

          calcium-binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA strand renaturation
          neutrophil homeostasis
          signal transduction
          cell surface receptor signaling pathway
          response to hormone
          response to X-ray
          response to organic cyclic compound
          peptide cross-linking
          insulin secretion
          keratinocyte differentiation
          prostate gland development
          endocrine pancreas development
          positive regulation of vesicle fusion
          positive regulation of prostaglandin biosynthetic process
          response to corticosteroid
          response to estradiol
          DNA duplex unwinding
          positive regulation of neutrophil apoptotic process
          DNA rewinding
          gliogenesis
          regulation of cell proliferation
          response to drug
          positive regulation of apoptotic process
          response to peptide hormone
          estrous cycle
          alpha-beta T cell differentiation
          arachidonic acid secretion
          negative regulation of protein secretion
          response to glucocorticoid
          cellular response to hydrogen peroxide
          hepatocyte differentiation
          prolactin secretion
          response to interleukin-1
          cellular response to glucocorticoid stimulus
          neutrophil clearance
          positive regulation of G1/S transition of mitotic cell cycle
          negative regulation of phospholipase A2 activity
          negative regulation of interleukin-8 secretion
          single-stranded DNA binding
          single-stranded RNA binding
          helicase activity
          structural molecule activity
          calcium ion binding
          phospholipid binding
          calcium-dependent phospholipid binding
          phospholipase A2 inhibitor activity
          protein binding, bridging
          double-stranded DNA-dependent ATPase activity
          annealing helicase activity
          protein homodimerization activity
          calcium-dependent protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P11614 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 479574 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618284 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 499841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 354791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 524820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869553 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 437811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78053 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 877739 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 180776 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 87075 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11614 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11614 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P11614 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P524359 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11614 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97243 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 96266 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10729281 - -
          Affymetrix Rat Gene 1.0 ST Array 10729275 - -
          Affymetrix Rat Gene 1.0 ST Array 10729277 - -
          Affymetrix Rat Gene 1.0 ST Array 10729270 - -
          Affymetrix Rat Gene 1.0 ST Array 10729276 - -
          Affymetrix Rat Gene 1.0 ST Array 10729279 - -
          Affymetrix Rat Gene 1.0 ST Array 10729272 - -
          Affymetrix Rat Gene 1.0 ST Array 10729274 - -
          Affymetrix Rat Gene 1.0 ST Array 10729278 - -
          Affymetrix Rat Gene 1.0 ST Array 10729280 - -
          Affymetrix Rat Gene 1.0 ST Array 10729273 - -
          Affymetrix Rat Gene 1.0 ST Array 10729271 - -
          Affymetrix Rat Gene 1.1 ST Array 10729269 - -
          Rat Genome 230 2.0 Array 1394451_at - -
          Rat Genome 230 2.0 Array 1367614_at - -
          Rat Genome 230 2.0 Array 1394451_PM_at - -
          Rat Genome 230 2.0 Array 1367614_PM_at - -
          Affymetrix HT Rat Focus Array 1367614_at - -
          Affymetrix HT Rat Focus Array 1394451_at - -
          Affymetrix HT Rat Focus Array 1394451_PM_at - -
          Affymetrix HT Rat Focus Array 1367614_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100059688.rn.1 TC0100007387.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100059685.rn.1 TC0100007387.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100059686.rn.1 TC0100007387.rn.1 RTA TC
          Rat Expression Set 230 1367614_PM_at - -
          Rat Expression Set 230 1367614_at - -
          Affymetrix Rat Gene 1.0 ST Array 10729269 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20845281 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353924 - -
          RatRef-12 Expression BeadChip ILMN_60211 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline