Search
Search

RGD1565775| Gene Symbol: RGD1565775 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_342277.6 | XP_342278.5 | 1 - 2 | 90 | 55 | |
| GenBank mRNA | BC092205.1 | - | 1 - 2 | 43 | 55 | |
| BC103646.1 | - | 1 - 2 | 68 | 55 | ||
Set Membership: |
|