Search
Search

RGD1565775| Gene Symbol: RGD1565775 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_342277.6 | XP_342278.5 | 2 - 3 | 302 | 93 | |
| GenBank mRNA | BC092205.1 | - | 2 - 3 | 255 | 93 | |
| BC103646.1 | - | 2 - 3 | 280 | 93 | ||
Set Membership: |
|