Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01455798_m1
          See other SSMEM1 GE Assays ›
          Gene Symbol:
          Ssmem1
          Gene Name:
          serine-rich single-pass membrane protein 1
          Gene Aliases:
          RGD1559502
          Chromosome Location:
          Chr.4: 57823411 - 57833706 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01455798_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ssmem1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001109228.1 NP_001102698.1 2 - 3 525 68
          XM_006236228.2 XP_006236290.1 2 - 3 477 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400000548.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P729029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 15317 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 845685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 49245 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153243 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906919 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602467 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158382 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 145478 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P729029 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P729029 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P729029 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P729029 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42007 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10854290 - -
          Affymetrix Rat Gene 1.0 ST Array 10854287 - -
          Affymetrix Rat Gene 1.0 ST Array 10854289 - -
          Affymetrix Rat Gene 1.1 ST Array 10854286 - -
          Affymetrix Rat Clariom S Assay PSR0400003915.rn.1 TC0400000548.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400003919.rn.1 TC0400000548.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400003917.rn.1 TC0400000548.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400003918.rn.1 TC0400000548.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400003916.rn.1 TC0400000548.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10854286 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20729193 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1374699 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline