Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01456549_m1
          See other STAT3 GE Assays ›
          Gene Symbol:
          Stat3
          Gene Name:
          signal transducer and activator of transcription 3 (acute-phase response factor)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 88790401 - 88842257 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01456549_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Stat3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012747.2 NP_036879.1 1 - 2 189 58
          XM_006247257.2 XP_006247319.1 1 - 2 213 58
          XM_006247258.2 XP_006247320.1 1 - 2 213 58
          XM_006247259.2 XP_006247321.1 1 - 2 213 58
          GenBank mRNA BC087025.1 - 1 - 2 189 7110655 58

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the BC087025.1, NM_012747.2, XM_006247257.2, XM_006247258.2, XM_006247259.2 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Rattus norvegicus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Stat3

          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          temperature homeostasis
          eye photoreceptor cell differentiation
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          protein import into nucleus
          acute-phase response
          JAK-STAT cascade
          aging
          cell proliferation
          negative regulation of cell proliferation
          response to organic substance
          negative regulation of hydrogen peroxide biosynthetic process
          response to organic cyclic compound
          phosphorylation
          stem cell population maintenance
          sexual reproduction
          intracellular receptor signaling pathway
          response to estradiol
          cellular response to hormone stimulus
          response to cytokine
          regulation of multicellular organism growth
          response to drug
          glucose homeostasis
          eating behavior
          response to peptide hormone
          response to ethanol
          positive regulation of Notch signaling pathway
          negative regulation of glycolytic process
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          regulation of mitochondrial membrane permeability
          astrocyte differentiation
          radial glial cell differentiation
          growth hormone receptor signaling pathway
          JAK-STAT cascade involved in growth hormone signaling pathway
          negative regulation of cell death
          interleukin-6-mediated signaling pathway
          negative regulation of neuron death
          positive regulation of growth factor dependent skeletal muscle satellite cell proliferation
          positive regulation of ATP biosynthetic process
          negative regulation of neuron migration
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          RNA polymerase II repressing transcription factor binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          signal transducer activity
          RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
          transcription factor binding
          protein kinase binding
          protein phosphatase binding
          CCR5 chemokine receptor binding
          glucocorticoid receptor binding
          sequence-specific DNA binding
          transcription regulatory region DNA binding
          protein dimerization activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Clariom S Assay PSR1000034288.rn.1 TC1000003816.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000034289.rn.1 TC1000003816.rn.1 RTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline