Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01456767_m1
          See other SPNS3 GE Assays ›
          Gene Symbol:
          Spns3
          Gene Name:
          spinster homolog 3 (Drosophila)
          Gene Aliases:
          RGD1561884
          Chromosome Location:
          Chr.10: 59056460 - 59113312 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01456767_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Spns3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001109035.1 NP_001102505.1 10 - 11 1487 80
          XM_006246786.2 XP_006246848.1 11 - 12 2076 80
          XM_006246789.2 XP_006246851.1 10 - 11 1924 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000003181.rn.1

          Set Membership:

          Amplicon length between 71 and 85 Transporter 3' Most Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          sphingosine-1-phosphate signaling pathway
          lipid transport
          locomotion
          transmembrane transport
          sphingolipid transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P014888 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 328094 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 332283 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 742480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 554936 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 689681 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 230387 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 742865 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 876322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363419 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347599 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662641 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 902330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 873434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720641 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 100818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622303 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505801 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843143 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698273 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912801 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 867918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273867 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827004 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781977 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142140 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 573527 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 852037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 568958 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 777270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 200294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 353312 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 819417 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 390754 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 845516 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 122541 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 9277 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 146133 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 36247 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 180112 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117581 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 97213 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P014888 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P014888 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P014888 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 44662 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10744738 - -
          Affymetrix Rat Gene 1.0 ST Array 10744743 - -
          Affymetrix Rat Gene 1.0 ST Array 10744739 - -
          Affymetrix Rat Gene 1.0 ST Array 10744747 - -
          Affymetrix Rat Gene 1.0 ST Array 10744745 - -
          Affymetrix Rat Gene 1.0 ST Array 10744750 - -
          Affymetrix Rat Gene 1.0 ST Array 10744740 - -
          Affymetrix Rat Gene 1.0 ST Array 10744744 - -
          Affymetrix Rat Gene 1.0 ST Array 10744741 - -
          Affymetrix Rat Gene 1.0 ST Array 10744742 - -
          Affymetrix Rat Gene 1.0 ST Array 10744746 - -
          Affymetrix Rat Gene 1.0 ST Array 10744737 - -
          Affymetrix Rat Gene 1.0 ST Array 10744749 - -
          Affymetrix Rat Gene 1.1 ST Array 10744736 - -
          Affymetrix Rat Clariom S Assay PSR1000028705.rn.1 TC1000003181.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10744736 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21942671 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364249 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline