Search
Search
| Gene Symbol: Ntrk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021589.1 | NP_067600.1 | 11 - 12 | 1427 | 125 | |
| XM_006232746.2 | XP_006232808.1 | 10 - 11 | 1400 | 125 | ||
| GenBank mRNA | M85214.1 | - | 11 - 12 | 1427 | 125 | |
Set Membership: |
|