Search
Search
| Gene Symbol: Tdo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022403.2 | NP_071798.1 | 10 - 11 | 1017 | 83 | |
| GenBank mRNA | BC089802.1 | - | 10 - 11 | 1013 | 7370033 | 83 |
| M55167.1 | - | 10 - 11 | 1017 | 83 | ||
Set Membership: |
|