Search
Search

Fgfr1op2| Gene Symbol: Fgfr1op2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201421.1 | NP_958824.1 | 4 - 5 | 718 | 80 | |
| XM_008763402.1 | XP_008761624.1 | 4 - 5 | 697 | 80 | ||
| GenBank mRNA | AY426740.1 | - | 4 - 5 | 718 | 80 | |
Set Membership: |
|