Search
Search
| Gene Symbol: Clec4d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003707.1 | NP_001003707.1 | 3 - 4 | 256 | 135 | |
| GenBank mRNA | AY339882.1 | - | 3 - 4 | 256 | 135 | |
| AY494065.1 | - | 3 - 4 | 256 | 135 | ||
Set Membership: |
|