Search
Search
| Gene Symbol: Arl5c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109046.1 | NP_001102516.1 | 5 - 6 | 543 | 64 | |
| XM_006247396.2 | XP_006247458.1 | 5 - 6 | 736 | 64 | ||
| XM_006247397.2 | XP_006247459.1 | 5 - 6 | 730 | 64 | ||
Set Membership: |
|