Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01460500_g1
          See other LOC497899 GE Assays ›
          Gene Symbol:
          LOC497899
          Gene Name:
          similar to hypothetical protein 4930503F14
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 36709018 - 36717981 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01460500_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC497899
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001017472.1 NP_001017472.1 4 - 5 691 119
          XM_006246312.1 XP_006246374.1 4 - 5 759 119
          XM_006246313.2 XP_006246375.1 4 - 5 1334 119
          XM_008767700.1 XP_008765922.1 4 - 5 1222 119
          XM_008767701.1 XP_008765923.1 4 - 5 843 119
          GenBank mRNA BC079296.1 - 4 - 5 691 7134494 119

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000002737.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene LOC497899

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P951676 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 704086 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 661199 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 971003 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 61193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 611591 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150049 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 172927 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P951676 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P951676 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P951676 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P951676 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 987 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45026 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10742481 - -
          Affymetrix Rat Gene 1.0 ST Array 10742478 - -
          Affymetrix Rat Gene 1.0 ST Array 10742480 - -
          Affymetrix Rat Gene 1.0 ST Array 10742479 - -
          Affymetrix Rat Gene 1.0 ST Array 10742477 - -
          Affymetrix Rat Gene 1.1 ST Array 10742476 - -
          Affymetrix Rat Clariom S Assay PSR1000024107.rn.1 TC1000002737.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10742476 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22229249 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351260 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1530465 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline