Search
Search
| Gene Symbol: Aurkb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053749.1 | NP_446201.1 | 3 - 4 | 336 | 68 | |
| XM_006246569.2 | XP_006246631.1 | 3 - 4 | 354 | 68 | ||
| XM_006246570.2 | XP_006246632.1 | 2 - 3 | 241 | 68 | ||
| XM_008767757.1 | XP_008765979.1 | 3 - 4 | 251 | 68 | ||
| XM_008767758.1 | XP_008765980.1 | 3 - 4 | 197 | 68 | ||
| GenBank mRNA | BC097297.1 | - | 3 - 4 | 254 | 7454285 | 68 |
| D89731.1 | - | 3 - 4 | 336 | 68 | ||
Set Membership: |
|