Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01461208_g1
          See other ZFP207 GE Assays ›
          Gene Symbol:
          Zfp207
          Gene Name:
          zinc finger protein 207
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 67762970 - 67784854 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01461208_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp207
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001039020.1 NP_001034109.1 10 - 11 1353 88
          XM_006247001.1 XP_006247063.1 11 - 12 1507 88
          XM_006247002.1 XP_006247064.1 11 - 12 1507 88
          XM_008768060.1 XP_008766282.1 11 - 12 1507 88
          XM_008768061.1 XP_008766283.1 11 - 12 1507 88
          XM_008768062.1 XP_008766284.1 10 - 11 1459 88
          GenBank mRNA BC100269.1 - 10 - 11 1353 7387460 88
          FQ221862.1 - 8 - 9 1674 88

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          DNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          mitotic sister chromatid segregation
          mitotic spindle assembly checkpoint
          attachment of spindle microtubules to kinetochore
          protein stabilization
          regulation of chromosome segregation
          microtubule binding
          heparin binding
          poly(A) RNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P091886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769093 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 719588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912473 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 579374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 245481 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 732477 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542710 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 229233 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100490 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P091886 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P091886 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P091886 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P455078 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24887 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10736655 - -
          Affymetrix Rat Gene 1.0 ST Array 10736654 - -
          Affymetrix Rat Gene 1.0 ST Array 10736648 - -
          Affymetrix Rat Gene 1.0 ST Array 10736651 - -
          Affymetrix Rat Gene 1.0 ST Array 10736660 - -
          Affymetrix Rat Gene 1.0 ST Array 10736658 - -
          Affymetrix Rat Gene 1.0 ST Array 10736653 - -
          Affymetrix Rat Gene 1.0 ST Array 10736650 - -
          Affymetrix Rat Gene 1.0 ST Array 10736657 - -
          Affymetrix Rat Gene 1.0 ST Array 10736652 - -
          Affymetrix Rat Gene 1.0 ST Array 10736659 - -
          Affymetrix Rat Gene 1.0 ST Array 10736649 - -
          Affymetrix Rat Gene 1.1 ST Array 10736647 - -
          Rat Genome 230 2.0 Array 1394591_PM_at - -
          Rat Genome 230 2.0 Array 1380529_PM_at - -
          Rat Genome 230 2.0 Array 1394591_at - -
          Rat Genome 230 2.0 Array 1397132_PM_at - -
          Rat Genome 230 2.0 Array 1397132_at - -
          Rat Genome 230 2.0 Array 1380529_at - -
          Affymetrix HT Rat Focus Array 1394591_PM_at - -
          Affymetrix HT Rat Focus Array 1394591_at - -
          Affymetrix Rat Clariom S Assay PSR1000011084.rn.1 TC1000001208.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000011085.rn.1 TC1000001208.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000011086.rn.1 TC1000001208.rn.1 RTA TC
          Rat Expression Set 230 1394591_PM_at - -
          Rat Expression Set 230 1394591_at - -
          Rat Expression Set 230 1397132_at - -
          Rat Expression Set 230 1397132_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10736647 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22061784 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366749 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline