Search
Search
| Gene Symbol: Cdk12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033867.1 | NP_001029039.1 | 7 - 8 | 2720 | 76 | |
| NM_138916.2 | NP_620271.1 | 7 - 8 | 2720 | 76 | ||
| XM_006247250.2 | XP_006247312.1 | 7 - 8 | 3271 | 76 | ||
| XM_006247251.2 | XP_006247313.1 | 7 - 8 | 3274 | 76 | ||
| XM_006247253.2 | XP_006247315.1 | 7 - 8 | 3273 | 76 | ||
| GenBank mRNA | AY072294.1 | - | 7 - 8 | 2720 | 76 | |
| AY962568.1 | - | 7 - 8 | 2720 | 76 | ||
Set Membership: |
|