Search
Search

RGD1307100| Gene Symbol: RGD1307100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006232275.2 | XP_006232337.1 | 10 - 11 | 1188 | 112 | |
| XM_008760914.1 | XP_008759136.1 | 10 - 11 | 1188 | 112 | ||
| XM_008760915.1 | XP_008759137.1 | 10 - 11 | 1188 | 112 | ||
| XM_008760916.1 | XP_008759138.1 | 10 - 11 | 1188 | 112 | ||
| XM_008760917.1 | XP_008759139.1 | 10 - 11 | 1188 | 112 | ||
| XM_008760918.1 | XP_008759140.1 | 10 - 11 | 1188 | 112 | ||
| XM_008760919.1 | XP_008759141.1 | 10 - 11 | 1187 | 112 | ||
Set Membership: |
|