Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01462414_m1
          See other RASIP1 GE Assays ›
          Gene Symbol:
          Rasip1
          Gene Name:
          Ras interacting protein 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 101608995 - 101621648 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01462414_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rasip1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106261.2 NP_001099731.1 9 - 10 1917 94
          XM_006229030.2 XP_006229092.1 11 - 12 3002 94
          GenBank mRNA BC168876.1 - 9 - 10 1917 7135603 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100008042.rn.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s)

          angiogenesis
          vasculogenesis
          negative regulation of autophagy
          branching morphogenesis of an epithelial tube

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1039809 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 64710 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 320506 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 175333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 768607 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187299 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 561118 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99166 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 24058 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1039809 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1039809 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1039809 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1039809 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 13948 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10706918 - -
          Affymetrix Rat Gene 1.0 ST Array 10706914 - -
          Affymetrix Rat Gene 1.0 ST Array 10706916 - -
          Affymetrix Rat Gene 1.0 ST Array 10706919 - -
          Affymetrix Rat Gene 1.0 ST Array 10706917 - -
          Affymetrix Rat Gene 1.0 ST Array 10706920 - -
          Affymetrix Rat Gene 1.0 ST Array 10706921 - -
          Affymetrix Rat Gene 1.0 ST Array 10706913 - -
          Affymetrix Rat Gene 1.0 ST Array 10706915 - -
          Affymetrix Rat Gene 1.1 ST Array 10706909 - -
          Rat Genome 230 2.0 Array 1388970_PM_at - -
          Rat Genome 230 2.0 Array 1388970_at - -
          Rat Genome 230 2.0 Array 1398505_at - -
          Rat Genome 230 2.0 Array 1398505_PM_at - -
          Rat Genome 230 2.0 Array 1377965_PM_at - -
          Rat Genome 230 2.0 Array 1375694_at - -
          Rat Genome 230 2.0 Array 1377965_at - -
          Rat Genome 230 2.0 Array 1375694_PM_at - -
          Affymetrix HT Rat Focus Array 1388970_at - -
          Affymetrix HT Rat Focus Array 1398505_PM_at - -
          Affymetrix HT Rat Focus Array 1388970_PM_at - -
          Affymetrix HT Rat Focus Array 1398505_at - -
          Affymetrix HT Rat Focus Array 1377965_at - -
          Affymetrix HT Rat Focus Array 1375694_PM_at - -
          Affymetrix HT Rat Focus Array 1375694_at - -
          Affymetrix HT Rat Focus Array 1377965_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100065299.rn.1 TC0100008042.rn.1 RTA TC
          Rat Expression Set 230 1375694_PM_at - -
          Rat Expression Set 230 1375694_at - -
          Rat Expression Set 230 1388970_PM_at - -
          Rat Expression Set 230 1388970_at - -
          Rat Expression Set 230 1398505_PM_at - -
          Rat Expression Set 230 1377965_PM_at - -
          Rat Expression Set 230 1377965_at - -
          Rat Expression Set 230 1398505_at - -
          Affymetrix Rat Gene 1.0 ST Array 10706909 - -
          Rat Genome U34 Set rc_AA848597_at - -
          Rat Genome U34 Set rc_AI101509_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21558294 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1374098 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline