Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01462970_mH
          See other CLEC2G GE Assays ›
          Gene Symbol:
          Clec2g
          Gene Name:
          C-type lectin domain family 2, member G
          Gene Aliases:
          CLEC2D11, Clr-b, Clr11, Clrb, RGD1563148
          Chromosome Location:
          Chr.4: 162934195 - 162942891 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01462970_mH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Clec2g
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001048075.1 NP_001041540.1 4 - 5 481 93
          GenBank mRNA DQ168417.1 - 4 - 5 481 93
          DQ168419.1 - 4 - 5 481 93
          EF100690.1 - 4 - 5 481 93
          EU128748.1 - 4 - 5 487 93

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400001879.rn.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Rattus norvegicus Probe spans exons

          Gene Ontology Categories:

          Process(es)

          carbohydrate binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P138179 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 150872 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 232965 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 281507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 942690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439783 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 506830 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 176700 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P138179 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P138179 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P138179 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P391750 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95369 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10859109 - -
          Affymetrix Rat Gene 1.0 ST Array 10859115 - -
          Affymetrix Rat Gene 1.0 ST Array 10859112 - -
          Affymetrix Rat Gene 1.0 ST Array 10859116 - -
          Affymetrix Rat Gene 1.0 ST Array 10859111 - -
          Affymetrix Rat Gene 1.0 ST Array 10859114 - -
          Affymetrix Rat Gene 1.1 ST Array 10859108 - -
          Rat Genome 230 2.0 Array 1390024_at - -
          Rat Genome 230 2.0 Array 1390024_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400013633.rn.1 TC0400001879.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400013629.rn.1 TC0400001879.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10859108 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353111 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline