Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01463179_g1
          See other TSEN2 GE Assays ›
          Gene Symbol:
          Tsen2
          Gene Name:
          TSEN2 tRNA splicing endonuclease subunit
          Gene Aliases:
          RGD1309946
          Chromosome Location:
          Chr.4: 147455489 - 147490869 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01463179_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tsen2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001014057.1 NP_001014079.1 4 - 5 347 89
          XM_008763240.1 XP_008761462.1 4 - 5 370 89
          XM_008763241.1 XP_008761463.1 4 - 5 409 89
          XM_008763242.1 XP_008761464.1 4 - 5 371 89
          GenBank mRNA BC087631.1 - 4 - 5 347 7319546 89

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tsen2

          Panther Classification:

          Molecular Function -

          endodeoxyribonuclease

          Gene Ontology Categories:

          Function(s) Process(es)

          tRNA-type intron splice site recognition and cleavage
          tRNA splicing, via endonucleolytic cleavage and ligation
          mRNA processing
          RNA phosphodiester bond hydrolysis
          RNA phosphodiester bond hydrolysis, endonucleolytic
          tRNA-intron endonuclease activity
          nucleic acid binding
          lyase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P114638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 232793 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770646 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171341 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 510438 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 431121 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 684778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360121 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156453 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 430056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698990 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 876699 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 680637 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 49288 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 559273 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 519999 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 401233 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 395943 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 169454 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 86441 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517941 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 127436 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 685289 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 14547 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 39465 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 15037 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 128143 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115794 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117728 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P114638 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P114638 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P114638 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P114638 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35178 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 53742 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92332 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10858001 - -
          Affymetrix Rat Gene 1.0 ST Array 10858008 - -
          Affymetrix Rat Gene 1.0 ST Array 10858006 - -
          Affymetrix Rat Gene 1.0 ST Array 10858007 - -
          Affymetrix Rat Gene 1.0 ST Array 10858003 - -
          Affymetrix Rat Gene 1.0 ST Array 10858005 - -
          Affymetrix Rat Gene 1.0 ST Array 10858004 - -
          Affymetrix Rat Gene 1.0 ST Array 10858010 - -
          Affymetrix Rat Gene 1.0 ST Array 10858009 - -
          Affymetrix Rat Gene 1.0 ST Array 10858002 - -
          Affymetrix Rat Gene 1.0 ST Array 10858000 - -
          Affymetrix Rat Gene 1.1 ST Array 10857999 - -
          Rat Genome 230 2.0 Array 1383430_at - -
          Rat Genome 230 2.0 Array 1383430_PM_at - -
          Affymetrix HT Rat Focus Array 1383430_at - -
          Affymetrix HT Rat Focus Array 1383430_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400011445.rn.1 TC0400001604.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400011448.rn.1 TC0400001604.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400011455.rn.1 TC0400001604.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400011456.rn.1 TC0400001604.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400011443.rn.1 TC0400001604.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400011447.rn.1 TC0400001604.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400011452.rn.1 TC0400001604.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400011451.rn.1 TC0400001604.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400011453.rn.1 TC0400001604.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400011442.rn.1 TC0400001604.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400011444.rn.1 TC0400001604.rn.1 RTA TC
          Rat Expression Set 230 1383430_PM_at - -
          Rat Expression Set 230 1383430_at - -
          Affymetrix Rat Gene 1.0 ST Array 10857999 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21783679 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1650034 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline