Search
Search
| Gene Symbol: Igfbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004274.2 | NP_001004274.1 | 2 - 3 | 737 | 83 | |
| GenBank mRNA | AY686592.1 | - | 2 - 3 | 501 | 83 | |
| BC098750.1 | - | 2 - 3 | 737 | 7368649 | 83 | |
Set Membership: |
|