Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01464174_m1
          See other KRT12 GE Assays ›
          Gene Symbol:
          Krt12
          Gene Name:
          keratin 12, type I
          Gene Aliases:
          Ka12, Krt1-12
          Chromosome Location:
          Chr.10: 87328547 - 87336709 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01464174_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Krt12
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001008761.1 NP_001008761.1 7 - 8 1275 75
          XM_006247357.2 XP_006247419.1 7 - 8 2287 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000003743.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Krt12

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          epithelium development
          cornea development in camera-type eye
          molecular_function
          structural molecule activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P319322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 175854 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197389 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P319322 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P319322 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P319322 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P319322 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 47338 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10747101 - -
          Affymetrix Rat Gene 1.0 ST Array 10747103 - -
          Affymetrix Rat Gene 1.0 ST Array 10747096 - -
          Affymetrix Rat Gene 1.0 ST Array 10747095 - -
          Affymetrix Rat Gene 1.0 ST Array 10747098 - -
          Affymetrix Rat Gene 1.0 ST Array 10747102 - -
          Affymetrix Rat Gene 1.0 ST Array 10747097 - -
          Affymetrix Rat Gene 1.0 ST Array 10747100 - -
          Affymetrix Rat Gene 1.1 ST Array 10747094 - -
          Rat Genome 230 2.0 Array 1373010_PM_at - -
          Rat Genome 230 2.0 Array 1373010_at - -
          Affymetrix HT Rat Focus Array 1373010_PM_at - -
          Affymetrix HT Rat Focus Array 1373010_at - -
          Affymetrix Rat Clariom S Assay PSR1000033541.rn.1 TC1000003743.rn.1 RTA TC
          Rat Expression Set 230 1373010_PM_at - -
          Rat Expression Set 230 1373010_at - -
          Affymetrix Rat Gene 1.0 ST Array 10747094 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21090575 - -
          ABI Rat Genome Survey Microarray 21306033 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376425 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline