Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01464878_m1
          See other CCNJL GE Assays ›
          Gene Symbol:
          Ccnjl
          Gene Name:
          cyclin J-like
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 29122249 - 29181616 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01464878_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ccnjl
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001037773.4 NP_001032862.2 4 - 5 741 59
          XM_006246133.2 XP_006246195.1 5 - 6 1427 59
          GenBank mRNA BC099113.1 - 2 - 3 373 59

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the BC099113.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ccnjl

          Panther Classification:

          Molecular Function -

          kinase activator

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P081412 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 299960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945048 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 426573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 768393 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 804403 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 271438 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 757754 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 448200 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 508242 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 479539 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 902619 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 246871 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 516877 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643483 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 659780 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 65585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 965381 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598363 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960945 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 752212 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 683338 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 645767 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 372026 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 462812 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 275023 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 644918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 406624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 890886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 348538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 970063 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 113828 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 30624 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 114942 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 162024 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 30289 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75342 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86353 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P081412 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P081412 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P081412 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 69426 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45074 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10732894 - -
          Affymetrix Rat Gene 1.0 ST Array 10732897 - -
          Affymetrix Rat Gene 1.0 ST Array 10732898 - -
          Affymetrix Rat Gene 1.0 ST Array 10732895 - -
          Affymetrix Rat Gene 1.0 ST Array 10732899 - -
          Affymetrix Rat Gene 1.0 ST Array 10732896 - -
          Affymetrix Rat Gene 1.1 ST Array 10732893 - -
          Rat Genome 230 2.0 Array 1374277_PM_at - -
          Rat Genome 230 2.0 Array 1374277_at - -
          Affymetrix HT Rat Focus Array 1374277_at - -
          Affymetrix HT Rat Focus Array 1374277_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000003597.rn.1 TC1000000419.rn.1 RTA TC
          Rat Expression Set 230 1374277_PM_at - -
          Rat Expression Set 230 1374277_at - -
          Affymetrix Rat Gene 1.0 ST Array 10732893 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20862788 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351363 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline