Search
Search
| Gene Symbol: Sgca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107039.1 | NP_001100509.1 | 3 - 4 | 397 | 88 | |
| XM_006247191.2 | XP_006247253.1 | 2 - 3 | 221 | 88 | ||
| GenBank mRNA | FQ215532.1 | - | 2 - 3 | 183 | 88 | |
Set Membership: |
|