Search
Search

Atxn7l3| Gene Symbol: Atxn7l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006247472.2 | XP_006247534.1 | 5 - 6 | 1787 | 126 | |
| XM_008768225.1 | XP_008766447.1 | 5 - 6 | 1789 | 126 | ||
Set Membership: |
|