Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01465737_m1
          See other MED4 GE Assays ›
          Gene Symbol:
          Med4
          Gene Name:
          mediator complex subunit 4
          Gene Aliases:
          Vdrip
          Chromosome Location:
          Chr.15: 55406543 - 55416816 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01465737_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Med4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024256.1 NP_001019427.1 5 - 6 520 142
          GenBank mRNA BC093402.1 - 5 - 6 520 7389622 142
          FQ231796.1 - 3 - 4 206 142

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500000823.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Med4

          Panther Classification:

          Molecular Function -

          general transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          transcription initiation from RNA polymerase II promoter
          biological_process
          intracellular steroid hormone receptor signaling pathway
          androgen receptor signaling pathway
          positive regulation of transcription, DNA-templated
          RNA polymerase II transcription cofactor activity
          molecular_function
          transcription cofactor activity
          receptor activity
          thyroid hormone receptor binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P19089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 475251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 132852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 707651 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779263 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854928 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 134134 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 114994 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P19089 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P19089 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P19089 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P19089 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92164 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92850 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10781447 - -
          Affymetrix Rat Gene 1.0 ST Array 10781445 - -
          Affymetrix Rat Gene 1.0 ST Array 10781450 - -
          Affymetrix Rat Gene 1.0 ST Array 10781448 - -
          Affymetrix Rat Gene 1.0 ST Array 10781444 - -
          Affymetrix Rat Gene 1.0 ST Array 10781449 - -
          Affymetrix Rat Gene 1.0 ST Array 10781446 - -
          Affymetrix Rat Gene 1.1 ST Array 10781443 - -
          Rat Genome 230 2.0 Array 1376196_PM_a_at - -
          Rat Genome 230 2.0 Array 1376196_a_at - -
          Rat Genome 230 2.0 Array 1386253_at - -
          Rat Genome 230 2.0 Array 1386253_PM_at - -
          Affymetrix HT Rat Focus Array 1376196_a_at - -
          Affymetrix HT Rat Focus Array 1386253_at - -
          Affymetrix HT Rat Focus Array 1386253_PM_at - -
          Affymetrix HT Rat Focus Array 1376196_PM_a_at - -
          Affymetrix Rat Clariom S Assay PSR1500005653.rn.1 TC1500000823.rn.1 RTA TC
          Rat Expression Set 230 1376196_PM_a_at - -
          Rat Expression Set 230 1386253_PM_at - -
          Rat Expression Set 230 1386253_at - -
          Rat Expression Set 230 1376196_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10781443 - -
          Rat Genome U34 Set rc_AA800290_g_at - -
          Rat Genome U34 Set rc_AA800290_at - -
          Rat Genome U34 Set rc_AI071206_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357358 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline