Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01465786_m1
          See other DIS3 GE Assays ›
          Gene Symbol:
          Dis3
          Gene Name:
          DIS3 exosome endoribonuclease and 3'-5' exoribonuclease
          Gene Aliases:
          RGD1304646
          Chromosome Location:
          Chr.15: 83466330 - 83494132 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01465786_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dis3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001127483.1 NP_001120955.1 20 - 21 2867 161
          XM_006252391.2 XP_006252453.1 21 - 22 2728 161
          XM_008770869.1 XP_008769091.1 20 - 21 2802 161
          GenBank mRNA BC166824.1 - 20 - 21 2867 8370691 161
          FQ210067.1 - 20 - 21 2879 161

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500002259.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          exoribonuclease

          Gene Ontology Categories:

          Function(s) Process(es)

          rRNA catabolic process
          positive regulation of GTPase activity
          CUT catabolic process
          nucleic acid phosphodiester bond hydrolysis
          RNA phosphodiester bond hydrolysis, exonucleolytic
          3'-5'-exoribonuclease activity
          endonuclease activity
          guanyl-nucleotide exchange factor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P464042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584364 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 432614 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 864140 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 856542 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 931482 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 140025 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517927 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 318729 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 367007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 845121 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848103 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 732961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813419 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674548 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669894 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 106942 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130202 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 67369 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 133223 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P464042 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P464042 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P464042 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P464042 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10785573 - -
          Affymetrix Rat Gene 1.0 ST Array 10785574 - -
          Affymetrix Rat Gene 1.0 ST Array 10785578 - -
          Affymetrix Rat Gene 1.0 ST Array 10785577 - -
          Affymetrix Rat Gene 1.0 ST Array 10785566 - -
          Affymetrix Rat Gene 1.0 ST Array 10785581 - -
          Affymetrix Rat Gene 1.0 ST Array 10785575 - -
          Affymetrix Rat Gene 1.0 ST Array 10785584 - -
          Affymetrix Rat Gene 1.0 ST Array 10785567 - -
          Affymetrix Rat Gene 1.0 ST Array 10785583 - -
          Affymetrix Rat Gene 1.0 ST Array 10785568 - -
          Affymetrix Rat Gene 1.0 ST Array 10785572 - -
          Affymetrix Rat Gene 1.0 ST Array 10785571 - -
          Affymetrix Rat Gene 1.0 ST Array 10785570 - -
          Affymetrix Rat Gene 1.0 ST Array 10785582 - -
          Affymetrix Rat Gene 1.0 ST Array 10785576 - -
          Affymetrix Rat Gene 1.0 ST Array 10785569 - -
          Affymetrix Rat Gene 1.0 ST Array 10785579 - -
          Affymetrix Rat Gene 1.0 ST Array 10785580 - -
          Affymetrix Rat Gene 1.1 ST Array 10785565 - -
          Affymetrix Rat Clariom S Assay PSR1500015141.rn.1 TC1500002259.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500015143.rn.1 TC1500002259.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10785565 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21952868 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351540 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline