Search
Search
| Gene Symbol: Sucla2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108387.2 | NP_001101857.2 | 7 - 8 | 971 | 117 | |
| GenBank mRNA | BC166998.1 | - | 6 - 7 | 891 | 117 | |
| CF110427.1 | - | 3 - 4 | 397 | 117 | ||
| FQ216292.1 | - | 3 - 4 | 337 | 117 | ||
Set Membership: |
|