Search
Search
| Gene Symbol: Nmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_148891.2 | NP_683689.1 | 7 - 8 | 1256 | 122 | |
| GenBank mRNA | AJ492222.1 | - | 7 - 8 | 877 | 122 | |
| BC097277.1 | - | 7 - 8 | 1256 | 7457194 | 122 | |
Set Membership: |
|