Search
Search

Slitrk5| Gene Symbol: Slitrk5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107284.1 | NP_001100754.1 | 1 | 4786 | 80 | |
| XM_006252444.2 | XP_006252506.1 | 2 | 5391 | 80 | ||
| XM_006252445.2 | XP_006252507.1 | 2 | 5054 | 80 | ||
| XM_006252446.2 | XP_006252508.1 | 2 | 4864 | 80 | ||
| XM_006252447.2 | XP_006252509.1 | 2 | 5160 | 80 | ||
Set Membership: |
|