Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01468146_m1
          See other UBAC2 GE Assays ›
          Gene Symbol:
          Ubac2
          Gene Name:
          UBA domain containing 2
          Gene Aliases:
          Phgdhl1
          Chromosome Location:
          Chr.15: 108286379 - 108433531 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01468146_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ubac2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001034937.2 NP_001030109.2 8 - 9 1053 99
          XM_006252509.2 XP_006252571.1 7 - 8 1079 99
          GenBank mRNA BC105842.1 - 3 - 4 436 7455960 99

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500001197.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ubac2

          Gene Ontology Categories:

          Function(s)

          protein localization to endoplasmic reticulum

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P199077 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14975 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 140726 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289604 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 776293 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574211 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527013 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 213283 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 21271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 314090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 938494 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 819696 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963633 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 731677 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 140212 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 617700 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677708 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 625100 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 570097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 41722 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746172 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 530373 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 400847 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681720 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963776 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 385447 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 28033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 26982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36182 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 167387 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 732567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 478623 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 659044 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 321549 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 931816 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 972129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 137959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380070 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 520370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 444628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 83393 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935287 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 141963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 702152 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 21314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539563 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435755 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 9238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243874 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1610 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 752423 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511823 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 907101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 327153 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 777551 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 217618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697296 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 392992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 83000 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 293655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 89280 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 552934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 857413 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 898316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149816 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826498 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 766967 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 407949 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682226 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238035 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 313507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 9843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 874955 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 665921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828098 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 114746 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 175633 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 165120 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 106490 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 28577 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 17789 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 107208 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 41221 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150124 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 67358 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 119544 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 97200 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 19749 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P199077 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P199077 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P199077 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P199077 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P172029 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75668 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93238 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 12893 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 43719 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89246 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 60761 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 14843 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55889 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 82055 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 13725 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10782122 - -
          Affymetrix Rat Gene 1.0 ST Array 10782121 - -
          Affymetrix Rat Gene 1.0 ST Array 10782120 - -
          Affymetrix Rat Gene 1.0 ST Array 10782117 - -
          Affymetrix Rat Gene 1.0 ST Array 10782123 - -
          Affymetrix Rat Gene 1.1 ST Array 10782113 - -
          Rat Genome 230 2.0 Array 1371846_PM_at - -
          Rat Genome 230 2.0 Array 1385466_at - -
          Rat Genome 230 2.0 Array 1385466_PM_at - -
          Rat Genome 230 2.0 Array 1371846_at - -
          Affymetrix HT Rat Focus Array 1385466_at - -
          Affymetrix HT Rat Focus Array 1385466_PM_at - -
          Affymetrix HT Rat Focus Array 1371846_PM_at - -
          Affymetrix HT Rat Focus Array 1371846_at - -
          Affymetrix Rat Clariom S Assay PSR1500007478.rn.1 TC1500001197.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500007480.rn.1 TC1500001197.rn.1 RTA TC
          Rat Expression Set 230 1371846_at - -
          Rat Expression Set 230 1371846_PM_at - -
          Rat Expression Set 230 1385466_at - -
          Rat Expression Set 230 1385466_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10782113 - -
          Rat Genome U34 Set rc_AA874876_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22309600 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358486 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline